Safranine O, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: CI 50240

MQ200

S8884-100G S8884-25G
S8884-100GEA 3450 NOK
S8884-100G S8884-25G
Safranine O, Sigma-Aldrich®
Safranine O
Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Tetthet: 1.447 g/cm³ (22 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6
EINECS: 207-518-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dc0c31be4a55937a0b5a174dbed0c0bc902b4a8&vid=0a2ca33d8ae9a3b4dc6b34fd80a483a5b5b6257a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR