Soil Sampler Pro
Leverandør: Sampling Systems
1620A-01
SAMP1620A-01EA 3025 NOK
SAMP1620A-01
Soil Sampler Pro
Prøvetakere Prøvetakere til faststoff
The soil sampler pro is ideal for use with lawns, meadows, and light to medium soils.
  • Collect a fast, cross sectional soil sample
  • Robust stainless steel design
  • Includes a footrest and wide handles
  • Available from stock
  • Fast delivery
The soil sampler pro is great for taking fast cross sectional samples of soil. The robust design, including footrest and wide handles, allows users to use extra force to push down into the soil.