Disposable bailers
Leverandør: Sampling Systems
5040A-1000 5040W-1000
SAMP5040A-1000EA 6200 NOK
SAMP5040A-1000 SAMP5040W-1000
Disposable bailers
Prøvetakere Prøvetakere til engangsbruk
Disposable bailers designed for sampling from wells or storage silos.
Forpakning: Individually bagged.
NB: Not suitable for high density liquids.