Reusable bailer
Leverandør: Sampling Systems
5075A-350
SAMP5075A-350EA 14250 NOK
SAMP5075A-350
Reusable bailer
Prøvetakere Væskeprøvetakere
Reusable bailer for sampling wells, ground water, tanks and bores.
  • Easy to clean
  • Simply lower to take the sample
  • Manufactured from chemically resistant FEP (similar to PTFE)
Primarily used in environmental and chemical sampling.