Liquid Dip Samplers
Leverandør: Sampling Systems
5175A-100 5175A-50
SAMP5175A-100EA 6125 NOK
SAMP5175A-100 SAMP5175A-50
Liquid Dip Samplers
Prøvetakere Prøveøser
The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.
  • Made from high quality and chemically resistant 316L stainless steel
  • Different sizes available
  • Convenient hands-free emptying feature
Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.