Sack Clips
Leverandør: Sampling Systems
8010A-1 8010M-1
SAMP8010A-1EA 2925 NOK
SAMP8010A-1 SAMP8010M-1
Sack Clips
Klemmer Universalklemmer
Sack clips are designed for the easy closing and sealing of bags.
  • Enables quick and easy sealing of bags and sacks
  • Heavy-duty design
  • Available in a range of materials
  • Available from stock
  • Fast delivery
The plastic sack clip is perfect for closing and sealing a range of different bags and sacks. Simply insert the bag into the jaws of the clip and click shut.