Bowls
Leverandør: Sampling Systems
8017H-30 8017H-30S
SAMP8017H-30EA 1350 NOK
SAMP8017H-30 SAMP8017H-30S
Bowls
Skåler Laboratoriebrett/-skåler
Mixing bowls, made of HDPE, ideal for laboratories, production and small scale work.
  • Robust construction
  • Individually wrapped
  • Food grade, FDA grade material of construction, EU 10/2011 compliant
  • BSE/TSE free