Beger with handle, SteriWare®
Leverandør: Sampling Systems
8049H-2000S 8049H-2000
SAMP8049H-2000SEA 4975 NOK
SAMP8049H-2000S SAMP8049H-2000
Beger with handle, SteriWare®
Beger
HDPE, translucent.
  • Individually bagged
  • BSE/TSE-free
  • Full batch traceability
  • Non graduated
  • Available as gamma irradiated sterile (SAL 10-6)
Disposable beakers have a wide base which make them very stable. The handle is rounded to make it comfortable to hold.