Micro Spatula
Leverandør: Sampling Systems
A106-20
SAMPA106-20EA 930 NOK
SAMPA106-20
Micro Spatula
Spatler
Designed for measuring and sampling small amounts of powder.
  • Ideal for surface sampling and dispensing small quantities
  • Crevice-free and easily cleaned
  • Available from stock
  • Fast delivery
High-quality micro spatulas are excellent for a variety of applications, such as measuring, sampling, or dispensing small quantities of powder, granule, and cream. They are crevice-free and therefore easily cleaned.