Containers with Sloping Base
Leverandør: Sampling Systems
A136-10B A136-01A A136-01S A616-50 A616-100 A616-200
300-0642EA 3350 NOK
300-0642 SAMPA136-01A SAMPA136-01S SAMPA616-50 SAMPA616-100 SAMPA616-200
Containers with Sloping Base
Tønner, fat, trommer
These drums which incorporate a sloping base allow the container to drain completely.
  • Made from 316L stainless steel
  • Includes lid, clamp and silicone seal
  • Other seals available
  • Ferrule for easy emptying
  • Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.