Stainless Steel Shovels
Leverandør: Sampling Systems
A246-10
SAMPA246-10EA 7700 NOK
SAMPA246-10
Stainless Steel Shovels
Spader
This stainless shovel is perfect for all general purpose work.
  • Made from chemically resistant 316L stainless steel
  • Ergonomic handle
  • Highly polished finish for easy cleaning
Made from high quality corrosion-resistant 316L stainles steel, it is crevice-free and has a high polish finish so can be easily cleaned once used.