Scoops, long handled, PharmaScoop®
Leverandør: Sampling Systems
A396-1000
SAMPA396-1000EA 4025 NOK
SAMPA396-1000
Scoops, long handled, PharmaScoop®
Øser
The long handled PharmaScoop made from 316 stainless steel is ideal for use with large containers.
  • 600 mm long handle allows the operator to safely reach deep into containers.
  • Crevice-free
  • Radiused corners for easy cleaning
  • GMP correct
  • BSE/TSE-free
Body length: 235 mm
Handle length: 600 mm