X-ray and metal detectable spoons
Leverandør: Sampling Systems
M1006B-10
SAMPM1006B-10EA 150 NOK
SAMPM1006B-10
X-ray and metal detectable spoons
Skjeer
The X-ray and metal detectable spoons are ideal for use in food and pharmaceutical production and sampling. The spoons can be used with both powders and liquids. They are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder or liquid in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Available in 10 ml volume
  • Full batch traceability
To ensure the cleanliness of the spoons they are manufactured and packed inMore Product Information