X-ray and metal detectable long handle scoops
Leverandør: Sampling Systems
M1040B-100
SAMPM1040B-100EA 475 NOK
SAMPM1040B-100
X-ray and metal detectable long handle scoops
Øser
These long handled metal detectable scoops are perfect for both sampling and dispensing when working with powders and granules.
  • Robust PP construction gives excellent chemical resistance to many products
  • Supplied Individually bagged and produced in a cleanroom
  • Full batch traceability
The long handle and the bright blue colour make the scoops easy to spot if they get left behind. They are both X-ray and metal detectable.
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. The robust nature of these scoops meansMore Product Information