Pipette calibration balance, Cubis®
Leverandør: Sartorius Balances
MPS66S
SARBMPS66SEA 0 NOK
SARBMPS66S
Pipette calibration balance, Cubis®
Vekter
The perfectly balanced system designed to reconcile all the needs for accuracy, precision, speed and convenience in pipette calibration.
  • Motion control, quick opening of the draft shield for convenient and efficient pipetting
  • Integrated humidity sensor - controls humidity inside the weighing chamber
  • Suitable for use in industrial areas
  • Class B interference emission
This balance features white LED light for illumination of the full weighing area and red LED light for instant alert if climate parameters are out of tolerance.
Sertifiseringer: EN 61326-More Product Information