Speedcal Mobile Fast Calibration of Multi-Channel Pipettes
Leverandør: Sartorius Balances
SPEEDCAL-M12 SPEEDCAL-M8
SARBSPEEDCAL-M12EA 0 NOK
SARBSPEEDCAL-M12 SARBSPEEDCAL-M8
Speedcal Mobile Fast Calibration of Multi-Channel Pipettes
Vekter
The Speedcal Mobile system was developed to meet the specific requirements for calibrating multi-channel pipettes. Its innovative design and sophisticated technology simplify the entire calibration process. Parallel connection of up to 12 balances, which simultaneously determine the dispensed volumes from a multi-channel pipette, make Speedcal Mobile the fastest and most effective system on the market for this task. The integrated Web Service interface enables Speedcal Mobile to be connected to all pipette calibration software. Speedcal Mobile is suitable for long-term use in laboratories.