Wire weight, OIML F1
Leverandør: Sartorius Balances
YCW1131-00
SARBYCW1131-00EA 530 NOK
SARBYCW1131-00
Wire weight, OIML F1
Veielodd Kalibreringslodd
Accurate metrological weight with nominal mass value of 10 mg.
  • OIML class F1
  • Hardness: ca. 110 HB
  • Density of 7,950 kg/m³
  • High gloss polished surface
  • No laser markings
  • Without certificate
Sertifiseringer: Weights comply with the International Recommendation OIML-R111.