Filtration capsules, Sartofluor®
Leverandør: Sartorius
5185307T9-SS-A 5181307T4-OO-B 5185307T7-OO-A 5185307T9-OO-A 5185307T7-SS-A 5185307T9-SO-A 5185307T7-SO-A 5181307T4-SO-B
SARP5185307T9-SS-AEA 16125 NOK
SARP5185307T9-SS-A SARP5181307T4-OO-B SARP5185307T7-OO-A SARP5185307T9-OO-A SARP5185307T7-SS-A SART5185307T9-SO-A SARP5185307T7-SO-A SARP5181307T4-SO-B
Filtration capsules, Sartofluor®
Filter Membranfilterkapsler
Sartofluor® MidiCaps® and Capsules are self contained, ready to use, sterile filter units for sterilising grade filtration in the pharmaceutical and biotechnology industry. Their unique hydrophobic PTFE membrane is ideally suited for particle removal and sterilising grade filtration of gases and for filtration of highly aggressive liquids like solvents, acids and bases.
  • Easy to use
  • Flexible and cost saving
  • Excellent flow rates for gases and liquids at low differential pressure
PTFE membrane; support fleece, core and end caps: PP; O-rings: EPDM;More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Inngang/utgang SS (1 ¹/₂" Sanitary flange/1 ¹/₂" Sanitary flange) OO (Hose barb/Hose barb) OO (Hose barb/Hose Barb) OO (Hose barb/Hose Barb) SS (1 ¹/₂" Sanitary flange/1 ¹/₂" Sanitary flange) SO (1 ¹/₂ '' Triclamp/Hose barb) SO (1 ¹/₂ '' Triclamp/Hose barb) SO (1 ¹/₂'' Triclamp/Hose barb)
Ø×L 77×197 mm 77×115 mm 77×197 mm 77×115 mm 77×197 mm 77×115 mm
Maks. trykk 5 bar @ 20 °C 4 bar @ 20 °C 5 bar @ 20 °C 5 bar @ 20 °C 5 bar @ 20 °C 5 bar @ 20 °C 5 bar @ 20 °C 4 bar @ 20 °C