Cellulose Extraction Thimbles, C300
Leverandør: Sartorius
FT-1201-030080 FT-1201-030100 FT-1201-033100 FT-1201-022100 FT-1201-025060 FT-1201-025070 FT-1201-028060 FT-1201-033060 FT-1201-033130 FT-1201-035150 FT-1201-040100 FT-1201-040123
SARTFT-1201-030080EA 1650 NOK
SARTFT-1201-030080 SARTFT-1201-030100 SARTFT-1201-033100 SARTFT-1201-022100 SARTFT-1201-025060 SARTFT-1201-025070 SARTFT-1201-028060 SARTFT-1201-033060 SARTFT-1201-033130 537- SARTFT-1201-040100 SARTFT-1201-040123
Cellulose Extraction Thimbles, C300
Ekstraktorer Ekstraksjonshylser
These cellulose thimbles are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Nr. C300
Materiale Cellulose