Kromatografipapir av cellulose

Leverandør: Sartorius
FT1-5270190100 FT2-527580600K FT2-527460570N FT2-527460570K FT-2503300580N FT-2507580600K FT-2527200200N FT2504-580600N FT2503-250250N FT2503-580600N FT-3-526-025
SARTFT1-5270190100EA 5325 NOK
SARTFT1-5270190100 SARTFT2-527580600K SARTFT2-527460570N 552-0374 SARTFT-2503300580N SARTFT-2507580600K 552-0373 SARTFT2504-580600N SARTFT2503-250250N SARTFT2503-580600N SARTFT-3-526-025
Kromatografipapir av cellulose
Filter Filterpapir Kromatografipapir
Chromatography papers are not only ideal for chromatography, but also for routine and repetitive separations, routine analysis of proteins in serum (e.g. human albumin) and several absorption applications like those common in the life sciences and diagnostics. They are made of cotton linters with more than 98% alpha-cellulose content and <0,04% ash content.
Bestill nå

SPESIFIKASJONER

Nr. FN 100 FN 3 FN 7 FN 4 FN 3 FN 30
Environmentally Preferable
Tykkelse 0,35 mm 0,19 mm 0,32 mm 0,24 mm 0,19 mm 0,9 mm
Vekt 120 g/m² 90 g/m² 320 g/m²
Kappilær stige 115 mm/30 min 95 mm/30 min 145 mm/30 min 95 mm/30 min 95 mm/30 min 240 mm/30 min

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR