Tork S4 Antimicrobial Hand Washing Liquid Soap
Leverandør: Essity
424709
SCAA424709EA 2225 NOK
SCAA424709
Tork S4 Antimicrobial Hand Washing Liquid Soap
Hudpleie Såper og hud rensemidler
Tork antimicrobial hand washing liquid soap S4, dermatologically tested.
  • Help to protect guests - this liquid soap conforms with EN 1499 and EN 14476 and is effective against all enveloped viruses
  • Prove your environmental commitment - this biodegradable liquid soap contains a 100% plant-based active ingredient
  • Be kind to skin - It is dermatologically tested and suitable for frequent use
  • Meet safety standards - this non-flammable soap has no CLP symbols, warning phrases or transport restrictions
  • No scent fragrance, premium quality
  • Colour: Transparent
More Product Information
New Product