Tork Reflex™ Wiping Paper
Leverandør: Essity
473242
SCAA473242EA 2250 NOK
SCAA473242
Tork Reflex™ Wiping Paper
Papirservietter Tørkepapir og våtservietter Papirservietter til generell bruk
The 1-ply multipurpose Tork Reflex™ Wiping Paper is ideal for wiping tasks and hand wiping, due to its strength and high absorption. This paper can be used in the Tork Reflex™ single-sheet centerfeed dispenser, which fits your team’s workflow, cuts down consumption, and minimises the risk of crosscontamination.
  • Single sheet dispensing - reduces consumption by up to 37%
  • SmartCore® core removal - for fast and easy refilling
  • Particularly good for cleaning glass - does not leave any lint or streaks on surfaces
System: M4 - Centrefeed Reflex roll system