Tork Clarity Foam Soap
Leverandør: Essity
520201
SCAA520201EA 1975 NOK
SCAA520201
Tork Clarity Foam Soap
Hudpleie Såper og hud rensemidler
Tork clarity foam soap contains sustainble formula - 99% of ingredients are of natural origin.
  • 35% lower water consumption* compared to Tork Mild Foam Soap (*Based on 2021 third-party verified LCA)
  • Washing ingredients are from renewable sources (vegetable origins)
  • The formula is dermatologically tested, of a skin-friendly pH and readily biodegradable (based on third-party lab test according to OECD301B)
  • Perfume free fragrance, premium quality
  • Colour: Transparent
  • Compatible with S4 dispensers
Sustainability aspirations can be achieved without