ThermoSafe® ChillTherm® Pre-Qualified 2 to 8 °C Shippers
Leverandør: Sonoco Thermosafe
CTR33
SCATCTR33EA 14425 NOK
SCATCTR33
ThermoSafe® ChillTherm® Pre-Qualified 2 to 8 °C Shippers
Forsending og transport kontainere
ChillTherm® pre-qualified 2 to 8 °C shippers are a seasonal, water-based solution which provides a cost-effective solution for the shipping of temperature-sensitive products.
  • Pre-qualified 2 to 8 °C shipper
  • Tested to several ambient profiles to speed up implementation
ChillTherm® shippers provide an economic solution when shipping products which need to be kept between 2 and 8 °C. Ideal for pharmaceuticals, biologics, medical devices, or animal health products, these shippers are easy to condition and pack out. Reports readily available so no need forMore Product Information