KIMBLE® KIMAX® Non-Mercury Thermometer for Pycnometer, 14 to 38 °C
Leverandør: DWK Life Sciences
15123N-2538
SCER15123N-2538EA 2975 NOK
SCER15123N-2538
KIMBLE® KIMAX® Non-Mercury Thermometer for Pycnometer, 14 to 38 °C
Termometere Analogue Thermometers
This non-mercury thermometer is designed for use in combination with a specific gravity pycnometer (10 or 25 ml).
  • Manufactured from borosilicate glass 3,3
  • Does not contain mercury
  • Features the same subdivisions, immersion depth, and precision as mercury-filled thermometers
  • Safer and more environmentally friendly than mercury-filled thermometers
Medium length standard taper ¹⁰/₁₈ thermometer with a temperature range of 14 to 38 °C. Accurate to ±0,5 °C.