Culture tubes, screw thread
Leverandør: DWK Life Sciences
D61251600
SCERD61251600EA 3250 NOK
SCERD61251600
Culture tubes, screw thread
Rør Kulturrør
Tubes, made up of borosilicate glass, grade 3.3, with screw threads.
  • Caps must be ordered separately
  • White marking spot
  • Very good chemical and temperature resistance