Volumetric flasks, class A, glass, ST ground glass stopper, KIMBLE®
Leverandør: DWK Life Sciences
SP28046-900
SCERSP28046-900EA 5125 NOK
SCERSP28046-900
Volumetric flasks, class A, glass, ST ground glass stopper, KIMBLE®
Kolber Målekolber
These volumetric flasks are calibrated both 'to contain' and 'to deliver' (top line) and designed from ASTM specification E288, Class A unserialized requirements.
  • Save shelf space, requires less space than a round flask (25 to 30%)
  • Graduation rings and marking spot are blasted onto glass