Culture tubes, disposable, KIMBLE®
Leverandør: DWK Life Sciences
SP60GM166
SCERSP60GM166EA 2600 NOK
SCERSP60GM166
Culture tubes, disposable, KIMBLE®
Rør Kulturrør
Manufactured from 90 expansion soda-lime glass, KIMAX®.
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths.
Sertifiseringer: Glass conforming USP Type III requirements