Screw caps for culture tubes, KIMBLE®
Leverandør: DWK Life Sciences
SP73805B-15415
SCERSP73805B-15415EA 2025 NOK
SCERSP73805B-15415
Screw caps for culture tubes, KIMBLE®
Lokk Lokk til reagensglass
PP.
  • Very good chemical resistance
  • Very good temperature resistance