Homogenisator, engangs, polypropylen, KIMBLE®
Leverandør: DWK Life Sciences
SP749540-0000
SCERSP749540-0000EA 4375 NOK
SCERSP749540-0000
Homogenisator, engangs, polypropylen, KIMBLE®
Homogenisatorer Homogenisatorer, håndholdt
Stainless steel cordless drive unit. Pressure is 14.7 PSI.
  • Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  • Operates at 2000 to 3000 min⁻¹ with fresh batteries and no load
Leveringsinformasjon: Supplied with 2× AA batteries.