KIMBLE® KIMAX® Cassia Volumetric Flask, Class A, TC, with Pennyhead Glass Stopper
Leverandør: DWK Life Sciences
SSP28066-110
SCERSSP28066-110EA 7450 NOK
SCERSSP28066-110
KIMBLE® KIMAX® Cassia Volumetric Flask, Class A, TC, with Pennyhead Glass Stopper
Kolber Målekolber
The KIMBLE® KIMAX® Cassia Volumetric Flask used in assay of oils of cinnamon, chenopodium, cloves, spearmint and other essential oils, according to methods of the United States Pharmacopoeia and the Essential Oil Association of the USA.
  • Manufactured from borosilicate glass 3,3
  • Calibrated 'To Contain'
  • Flask neck is graduated from 100 to 110 ml in 0,1 ml subdivisions, with a ring at each 1ml
  • With a marking spot and a duarbale white ceramic enamel scale
  • ASTM Class A
Supplied with pennyhead glass stopper (850100-0013).