Allergen lateral flow IIR
Leverandør: HYSERVE
1500001 1500002 1500004
SCIL1500001EA 3900 NOK
SCIL1500001 SCIL1500002 SCIL1500004
Allergen lateral flow IIR
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Lateral Flow
To detect allergen in highly processed foods by special extraction solution and unique antibody. This lateral-flow test is a high-performance qualitative assay.
  • Easy to use and fast assay
  • Available for various samples
  • 20 sticks/kit