Biowave 3+ Colour Touch Spectrophotometer
Leverandør: BIOCHROM
80-3007-57
SCLI80-3007-57EA 0 NOK
SCLI80-3007-57
Biowave 3+ Colour Touch Spectrophotometer
Spektrofotometre og fotometre Biofotometere
The Biowave 3+ colour touch offers all the benefits and capabilities of a research grade life science spectrophotometer with the robust design, ease of use and reasonable cost that also makes it perfect for students.
  • Pre-loaded life science methods:
  • Nucleic Acid quantification
  • Protein applications (Bradford, Lowry, Biuret, BSA)
  • Nucleic Acid and Protein fluorescent dyes
  • OD 600 Microbial growth
  • Tm calculation
Fast and easy to use wavelength scanning, kinetics, single or multiple wavelength reading, standard curve and absorbance ratio capability.