Biochrom Ultrospec® 35c Colorimeter
Leverandør: BIOCHROM
80-3008-00
SCLI80-3008-00EA 29650 NOK
SCLI80-3008-00
Biochrom Ultrospec® 35c Colorimeter
Kolorimeter Multiparameter Kolorimeter
The Ultrospec 35c Colorimeter allows you to store data directly to the device and export it to a computer.
  • Flexible analysis: Measures samples in absorbance and % Transmission modes as well as having simple kinetics functionality
  • Robust LED light source: User-selectable wavelengths: 450, 500, 520, 540, 600 and 640 nm
  • Touchscreen display: The instrument has a 109,22 mm colour touchscreen for easy workflow
  • Instrument data output: Measurement data can be exported to a USB flash drive as comma-separated values (.csv)
Versatile and portable: The instrumentMore Product Information