Pocket thermometers
Leverandør: Ludwig Schneider GmbH & CO. KG
1021142
SCNL1021142EA 375 NOK
SCNL1021142
Pocket thermometers
Termometere Analogue Thermometers
These pocket thermometers are protected designs (DBGM). Mercury, red or blue filling.
  • Total immersion
  • Available in different capillary forms
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.