Transmitted darkfield light stage, VisiLED, TLS-DF
Leverandør: SCHOTT AG Lighting and imaging
400350
SCOC400350EA 13225 NOK
SCOC400350
Transmitted darkfield light stage, VisiLED, TLS-DF
Lys Kaldlyskilder
Professional incident and darkfield illumination.
  • The VisiLED darkfield backlight TLS-DF offers the possibility of transmitted darkfield illumination
  • Lifetime is typically 50000 h by actively controlling the LED‘s temperature
Designed for use with controller MC 750, MC 1100, or MC 1500.
Bestillingsinformasjon: Required controllers must be purchased seperately.