Polariser set for ring light, VisiLED, S40-75
Leverandør: SCHOTT AG Lighting and imaging
400575
SCOC400575EA 6200 NOK
SCOC400575
Polariser set for ring light, VisiLED, S40-75
Lys Ring Lights
Polarisation system for brightfield ringlights. Turnable polariser with adapting thread and sliding filter as analyser.
  • Polarised light by a polarising and analysing filter within the beam path can be very efficiently used in stereo microscopy
  • Use to contrast inner material structure or to identify metal particles within others
Designed for use with VisiLED ring light S40-75.
Bestillingsinformasjon: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.