Diffusor for ring light, VisiLED, S40-75
Leverandør: SCHOTT AG Lighting and imaging
400580
SCOC400580EA 3650 NOK
SCOC400580
Diffusor for ring light, VisiLED, S40-75
Lys Ring Lights
The diffusor for LED ring lights avoids reflections of LEDs for uniform illumination.
  • Diffusor with adapting thread (M110×1) to avoid reflections of LEDs
Designed for use only with VisiLED ringlight S40-75.
Bestillingsinformasjon: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.