Automatic viscometer with sampler, AVS® Pro III
Leverandør: SI Analytics
285420030
SCOI285420030EA 0 NOK
SCOI285420030
Automatic viscometer with sampler, AVS® Pro III
Viskosimetere Kapillærviskosimeter
The AVS Pro III with autosampler has been developed for viscosity measurements in the polymer and petroleum industries.
  • High sample throughput
  • Process controlled by PC (not supplied) using AVSPro software (supplied)
  • ProClean system and micro dosing make routine operation safer particularly for aggressive sample fluids
  • Includes choice of two sample racks 16×100 ml or 56×20 ml
  • Optoelectronic and TC scanning for clear and opaque samples
  • Three axis-positioning sample dosing system
The X-Y-Z positioning mechanism provides parallel operation of up to eightMore Product Information