Sedimentation cones, Imhoff type, DURAN®
Leverandør: DWK Life Sciences
214005402
SCOT214005402EA 16250 NOK
SCOT214005402
Sedimentation cones, Imhoff type, DURAN®
Trakter Immhoff-trakter
DURAN®, borosilicate glass 3.3, clear.
  • Without graduations
  • Very good chemical resistance
  • Very good temperature resistance