DURAN® Blanks for Distillation Flasks, Engler Type
Leverandør: DWK Life Sciences
216512805
SCOT216512805EA 39375 NOK
SCOT216512805
DURAN® Blanks for Distillation Flasks, Engler Type
Kolber Destillasjonskolber
DURAN® borosilicate glass 3.3, clear.
  • With beaded rim
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.