Jars for organ storage, DURAN®
Leverandør: DWK Life Sciences
242042409
SCOT242042409EA 5300 NOK
SCOT242042409
Jars for organ storage, DURAN®
Krukker Prøveglass
DURAN®, borosilicate glass 3.3, clear. Without stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Organ storage jars, cylindrical vessels with a wide neck, are used to store materials.