Drying tubes with standard ground cone, bent, DURAN®
Leverandør: DWK Life Sciences
242620609 242620703 242620806 242620909
SCOT242620609EA 495 NOK
SCOT242620609 SCOT242620703 SCOT242620806 SCOT242620909
Drying tubes with standard ground cone, bent, DURAN®
Rør Tørkerør
DURAN® borosilicate glass 3.3. clear. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.