Distillation flasks, engler, with side arm, DURAN®
Leverandør: DWK Life Sciences
216542808 246542805
SCOT216542808EA 8300 NOK
SCOT216542808 SCOT246542805
Distillation flasks, engler, with side arm, DURAN®
Kolber Destillasjonskolber
These flasks are manufactured from a hydrolytically resistant borosilicate glass 3.3, which is defined as Type I, Class A in the ASTM Specification E 438.
  • Capacity: 125 ml
  • Side arm:100 mm long, 7 mm diameter
These flasks are available with two different fittings, either a NS 19/26 ground joint or a beaded rim.
Sertifiseringer: Comply with the requirements of ASTM D86 and DIN EN ISO 3405.