Desiccators, MOBILEX type with procelain plate, DURAN®
Leverandør: DWK Life Sciences
247835753 247836158 247836955 247836655
SCOT247835753EA 5150 NOK
SCOT247835753 SCOT247836158 SCOT247836955 SCOT247836655
Desiccators, MOBILEX type with procelain plate, DURAN®
Eksikatorer Eksikator, Glass
DURAN® clear borosilicate glass 3.3, vacuum desiccator with Mobilex screw thread in the lid with stopcock. With desiccator base with flat flange, no outlet, PBT screw cap with pierced aperture GL 32.
Leveringsinformasjon: Supplied with porclain plate and silicone sealing ring with bonded PTFE face