Stopcock keys for burettes, DURAN®
Leverandør: DWK Life Sciences
286035504
SCOT286035504EA 275 NOK
SCOT286035504
Stopcock keys for burettes, DURAN®
Stoppekran-nøkler
DURAN®, borosilicate glass 3.3, clear.
  • Side entry
  • Drilled and grounded
  • With three part retaining device