Stopcocks for burettes, straight, DURAN®
Leverandør: DWK Life Sciences
286449405
SCOT286449405EA 16000 NOK
SCOT286449405
Stopcocks for burettes, straight, DURAN®
Haner
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.