Stopcocks for burettes, angled, DURAN®
Leverandør: DWK Life Sciences
286451608
SCOT286451608EA 15375 NOK
SCOT286451608
Stopcocks for burettes, angled, DURAN®
Haner
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.