PTFE coated GL 45 high purity silicone septum, DURAN®
Leverandør: DWK Life Sciences
292483005
SCOT292483005EA 1375 NOK
SCOT292483005
PTFE coated GL 45 high purity silicone septum, DURAN®
Septum
A high purity PTFE coated silicone septum for use with DURAN® GL 45 open top screw caps and all sises of DURAN® GL 45 laboratory bottles. Can be used for sampling using a syringe and a needle.
  • Soft silicone rubber for easy needle penetration and resealability
  • Manufactured from high purity, Platinum catalysed silicone rubber
  • High quality PTFE film reduces the risk of solvent contamination
  • Wide temperature usage range from – 50 up to + 180°C
  • Suitable for autoclaving and dry heat sterilisation
Applications: Liquid transfer from HPLC solvent bottles,More Product Information