Water jet pump, DURAN®
Leverandør: DWK Life Sciences
292500101
SCOT292500101EA 1700 NOK
SCOT292500101
Water jet pump, DURAN®
Pumper Vannstrålepumper
Made of polypropylene.
  • Water consumption: 400 l/h at 3,5 bar flow pressure, at 12 °C
  • With non-return valve
  • With hose connection and adapters for 1/2" and 3/4"